Gold(I) iodide ≥99% (99.9+%-Au)

Supplier: STREM CHEMICALS INC MS
Warning

79-2000-5G 79-2000-1G
100195-924EA 1062.95 USD
100195-924 100206-844
Gold(I) iodide ≥99% (99.9+%-Au)
Gold(I) iodide
Formula: AuI
Melting Pt: 120 °C (dec.)
Density: 8.25
MDL Number: MFCD00014175
CAS Number: 10294-31-2
EINECS: 233-656-3
UN: 3260
ADR: 8,III
Merck Index: 12,04530

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR