tetra-Chloroauric(III) acid hydrate ≥99.8% (Au basis) (min. 49% Au)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Gold(III) chloride hydrate, Hydrogen tetrachloroaurate(III) xhydrate, Hydrogen tetrachloroaurate (III) hydrate, Hydrogen tetrachloroaurate hydrate, Chloroauric acid, tetra-Chloroauric acid hydrate

79-0500-25G 79-0500-5G 79-0500-1G
100198-910EA 2915.69 USD
100198-910 100196-396 100210-098
tetra-Chloroauric(III) acid hydrate ≥99.8% (Au basis) (min. 49% Au)
tetra-Chloroauric(III) acid hydrate
Formula: HAuCl₄·H₂O
MW: 357.8 g/mol
Density: 1.169 g/cm³ (20 °C)
MDL Number: MFCD00149903
CAS Number: 27988-77-8
EINECS: 240-948-4
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR