Disodium hexachloroplatinate hexahydrate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Sodium hexachloroplatinate(IV) hexahydrate, Sodium hexachloroplatinate (IV) hexahydrate

78-1995-5G 78-1995-1G
100203-824EA 751.57 USD
100203-824 100199-456
Disodium hexachloroplatinate hexahydrate ≥98%
Disodium hexachloroplatinate hexahydrate
Formula: Cl₆Na₂Pt·6H₂O
MW: 561.87 g/mol
Density: 2.5 g/cm³ (20 °C)
MDL Number: MFCD00149172
CAS Number: 19583-77-8
EINECS: 240-983-5
UN: 3288
ADR: 6.1,III
Merck Index: 12,08768

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR