Potassium tetrachloroplatinate(II) ≥99.9% (Pt basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
78-1970-5G
78-1970-25G
78-1970-1G
100199-994EA
714.77
USD
100199-994
100199-992
100208-476
Potassium tetrachloroplatinate(II) ≥99.9% (Pt basis)
Potassium tetrachloroplatinate(II)
Formula:
Cl₄K₂Pt Boiling Pt: 35…40 °C (20 mmHg) Melting Pt: 265 °C Density: 3.38 |
MDL Number:
MFCD00011378 CAS Number: 10025-99-7 EINECS: 233-050-9 UN: 3288 ADR: 6.1,III Merck Index: 12,07855 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...