Potassium hexachloroplatinate(IV) ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Dipotassium hexachloroplatinate
78-1960-5G
78-1960-1G
100196-394EA
755.82
USD
100196-394
100196-392
Potassium hexachloroplatinate(IV) ≥99%
Potassium hexachloroplatinate(IV)
Formula:
K₂PtCl₆ MW: 485.99 g/mol Melting Pt: 250 °C Density: 3.344 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011389 CAS Number: 16921-30-5 EINECS: 240-979-3 UN: 3290 ADR: 6.1,II Merck Index: 13,07720 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...