Platinum(II) chloride ≥99.9% (Pt basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Platinum dichloride

78-1480-5G 78-1480-1G
100196-384EA 1268.18 USD
100196-384 100196-382
Platinum(II) chloride ≥99.9% (Pt basis)
Platinum(II) chloride
Formula: Cl₂Pt
MW: 265.99 g/mol
Melting Pt: 581 (dec.)
Density: 6.05
MDL Number: MFCD00011181
CAS Number: 10025-65-7
EINECS: 233-034-1
UN: 3260
ADR: 8 III
Merck Index: 12,07687

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR