Platinum, black ≥97%

Supplier: STREM CHEMICALS INC MS

Synonyms: Platinium

78-1420-5G 78-1420-1G
100204-108EA 1279.51 USD
100204-108 100197-170
Platinum, black ≥97%
Platinum
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.45
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 3089
ADR: 4.1 III
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR