Platinum(II) acetylacetonate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

78-1400-1G 78-1400-5G
100201-750EA 157.11 USD
100201-750 100203-310
Platinum(II) acetylacetonate ≥98%
Platinum(II) acetylacetonate

Metal Beta-diketonates, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₀H₁₄O₄Pt
Melting Pt: 250 - 252 °C
Density: ~ 1 g/cm³ (20 °C)
MDL Number: MFCD00000028
CAS Number: 15170-57-7
EINECS: 239-223-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR