trans-Platin ≥99%
Supplier: STREM CHEMICALS INC MS
Synonyms:
trans-Diamminedichloroplatinum (II), trans-Diammineplatinum (II) dichloride, trans-Dichlorodiammine platinum(II), trans-Dichlorodiamineplatinum(II)
78-0451-1G
78-0451-5G
100198-036EA
324.12
USD
100198-036
100197-614
trans-Platin ≥99%
trans-Platin
Formula:
H₆Cl₂N₂Pt MW: 300.05 g/mol Melting Pt: 340 °C Density: 3.746 g/cm³ (20 °C) |
MDL Number:
MFCD00011623 CAS Number: 14913-33-8 EINECS: 238-980-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...