Platinum ≥99.95%, wire

Supplier: STREM CHEMICALS INC MS

Synonyms: Platinium

78-0085-100CM 78-0085-25CM
100209-274EA 1227.14 USD
100209-274 100208-462
Platinum ≥99.95%, wire
Platinum
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.45
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR