Iridium(III) chloride hydrate ≥99.9% (Ir basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Iridium trichloride hydrate, Iridium(III) chloride xhydrate
77-3000-5G
77-3000-1G
100205-198EA
2386.34
USD
100205-198
100205-196
Iridium(III) chloride hydrate ≥99.9% (Ir basis)
Iridium(III) chloride hydrate
Formula:
Cl₃Ir·xH₂O MW: 298.58 g/mol (anhy) Density: 5.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149750 CAS Number: 14996-61-3 EINECS: 233-044-6 Merck Index: 14,05088 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...