Diammonium hexachloroiridate ≥99%
Supplier: STREM CHEMICALS INC MS
77-0020-5G
77-0020-1G
100200-302EA
1314.89
USD
100200-302
100206-802
Diammonium hexachloroiridate ≥99%
Diammonium hexachloroiridate
Formula:
H₈Cl₆IrN₂ Melting Pt: Dec. Density: 2.85 g/cm³ (20 °C) |
MDL Number:
MFCD00010881 CAS Number: 16940-92-4 EINECS: 241-007-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...