Ammonium iron (III) sulfate dodecahydrate

Warning

F3629-250G F3629-500G
220177-270EA 22.91 USD
220177-270 220177-271
Ammonium iron (III) sulfate dodecahydrate
Ammonium iron(III) sulphate dodecahydrate
Formula: NH₄Fe(SO₄)₂·12H₂O
MW: 482.2 g/mol
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 39 - 41 °C
Density: 1.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150004
CAS Number: 7783-83-7
EINECS: 233-382-4
Merck Index: 12,00549

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR