Iron(III) chloride hexahydrate, Reagent Grade

Danger

F2877-100G F2877-500G F2877-250G F2877-1KG
220177-173EA 21.72 USD
220177-173 220177-176 220177-175 220177-174
Iron(III) chloride hexahydrate, Reagent Grade
Iron(III) chloride hexahydrate
Formula: FeCl₃·6H₂O
Boiling Pt: 280 - 285 °C (1013 hPa)
Melting Pt: 37 °C
Density: 1.82 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: 8,III
Merck Index: 13,04048
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=6d820873011bee3ff401441544f7f85599e40626&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR