EDTA dipotassium salt dihydrate

ED2P-500G ED2P-100G ED2P-1KG
220124-393EA 329.37 USD
220124-393 220124-391 220124-392
EDTA dipotassium salt dihydrate
EDTA dipotassium salt dihydrate
Formula: C₁₀H₁₄K₂N₂O₈·2H₂O
Melting Pt: 272 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150036
CAS Number: 25102-12-9
EINECS: 217-895-0
Merck Index: 13,03546

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR