Bis(2-ethoxyethyl)ether ≥98%, Reagent Grade

Synonyms: 2-Ethoxyethyl ether, Diethyleneglycoldiethylether, Diethylene glycol diethyl ether

E4658-500ML E4658-1L
220124-207EA 46.46 USD
220124-207 220122-674
Bis(2-ethoxyethyl)ether ≥98%, Reagent Grade
Bis(2-ethoxyethyl)ether
Formula: C₈H₁₈O₃
MW: 162.23 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: -44 °C
Density: 0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009254
CAS Number: 112-36-7
EINECS: 203-963-7
Merck Index: 13,03145

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR