Ethyl piperazine-1-carboxylate

Synonyms: 1-Ethoxycarbonyl piperazine, Ethyl N-piperazinecarboxylate

E45600-25G E45600-100G
220124-192EA 62.78 USD
220124-192 220124-191
Ethyl piperazine-1-carboxylate
Ethyl piperazine-1-carboxylate
Formula: C₇H₁₄N₂O₂
MW: 158.2 g/mol
MDL Number: MFCD00005964
CAS Number: 120-43-4
EINECS: 204-395-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR