EDTA tetrasodium salt hydrate ≥98%

Danger

E26290-25G E26290-500G E26290-10KG E26290-3KG
220123-958EA 33.38 USD
220123-958 220123-960 220123-957 220123-959
EDTA tetrasodium salt hydrate ≥98%
EDTA tetrasodium salt hydrate
Formula: C₁₀H₁₂N₂O₈Na₄·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00150025
CAS Number: 13235-36-4
EINECS: 200-573-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR