Ethyl 2-bromoisobutyrate ≥98%

Warning

Synonyms: Ethyl α-bromoisobutyrate, Ethyl 2-bromo-2-methylpropionate, 2-Brom-2-methylpropionic acid ethyl ester

E14403-100G E14403-500G E14403-5G
220123-762EA 58.91 USD
220123-762 220123-763 220123-764
Ethyl 2-bromoisobutyrate ≥98%
Ethyl 2-bromoisobutyrate
Formula: C₆H₁₁BrO₂
MW: 195.06 g/mol
Boiling Pt: 65 °C (1013 hPa)
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00000123
CAS Number: 600-00-0
EINECS: 209-980-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR