Diethylenetriamine ≥99%, ReagentPlus®

Danger

Synonyms: 2,2'-Iminodi(ethylamine), DIEN, Diethylenetriamine

D93856-1L D93856-18L D93856-5ML D93856-100ML
220123-359EA 60.23 USD
220123-359 220123-358 220123-361 220123-357
Diethylenetriamine ≥99%, ReagentPlus®
Diethylenetriamine
Formula: C₄H₁₃N₃
MW: 103.17 g/mol
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: –39 °C
Density: 0.9523 g/cm³ (20 °C)
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00008171
CAS Number: 111-40-0
EINECS: 203-865-4
UN: 2079
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR