Silver diethyldithiocarbamate ≥99% ACS

Warning

Synonyms: N,N-Diethyldithiocarbamic acid silver salt, Diethyldithiocarbamic acid silver salt, DETC, (Diethyldithiocarbamato-S,S')silver

D93503-25G D93503-5G
220123-349EA 270 USD
220123-349 220123-350
Silver diethyldithiocarbamate ≥99% ACS
Silver diethyldithiocarbamate
Formula: (C₂H₅)₂NCSSAg
MW: 256.14 g/mol
Melting Pt: 176 - 178.5 °C
MDL Number: MFCD00004929
CAS Number: 1470-61-7
EINECS: 216-003-7
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR