Diethanolamine ≥98%, Reagent Grade

Danger

D8885-1KG D8885-500G
220123-221EA 55.31 USD
220123-221 220123-222
Diethanolamine ≥98%, Reagent Grade
Diethanolamine
Formula: C₄H₁₁NO₂
Boiling Pt: 268.8 °C (1013 hPa)
Melting Pt: 28 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00002843
CAS Number: 111-42-2
EINECS: 203-868-0
REACH: 01-2119488930-28
Merck Index: 13,03134

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR