3,4-Dichlorobenzaldehyde ≥95%

Danger

D56608-5G D56608-25G
220122-135EA 42.43 USD
220122-135 220122-134
3,4-Dichlorobenzaldehyde ≥95%
3,4-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 248 °C (1013 hPa)
Melting Pt: 43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003351
CAS Number: 6287-38-3
EINECS: 228-520-5
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR