2,6-Di-tert-butylphenol ≥99%

Synonyms: 1,3-Di(dimethylethyl)-2-hydroxybenzene

D48400-500G D48400-25G D48400-1KG
220122-200EA 55.46 USD
220122-200 220122-199 220122-198
2,6-Di-tert-butylphenol ≥99%
2,6-Di-tert-butylphenol
Formula: C₁₄H₂₂O
MW: 206.33 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 34…37 °C
Density: 0.914 g/cm³ (20 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00008820
CAS Number: 128-39-2
EINECS: 204-884-0
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR