Dansyl chloride

Danger

Synonyms: 5-(Dimethylamino)naphthalene-1-sulphonyl chloride

D2625-5G D2625-1G D2625-25G
220121-724EA 437.31 USD
220121-724 220121-722 220121-723
Dansyl chloride
Dansyl chloride
Formula: C₁₂H₁₂ClNO₂S
MW: 269.75 g/mol
Melting Pt: 69 - 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003985
CAS Number: 605-65-2
EINECS: 210-092-6
UN: 3261
ADR: 8,III
Merck Index: 13,02840

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR