Decahydronaphthalene (mixture of cis and trans isomers) ≥98%, Reagent Grade

Danger

Synonyms: Decalin, Bicyclo[4.4.0]decane, Decahydronaphthalene

D251-4L D251-1L
220121-170EA 20.6 USD
220121-170 220121-169 220121-167
Decahydronaphthalene (mixture of cis and trans isomers) ≥98%, Reagent Grade
Decahydronaphthalene (mixture of cis and trans isomers)
Formula: C₁₀H₁₈
MW: 138.25 g/mol
Boiling Pt: 189 - 191 °C (1013 hPa)
Melting Pt: ~ -40 °C
Density: 0.8696 g/cm³ (20 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00004130
CAS Number: 91-17-8
EINECS: 202-046-9
UN: 1147
ADR: 3,III
Merck Index: 13,02866
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41b5fff8aa81be0e42af177a8f0e3ac149943c6e&vid=ca9f3c5d71ac621a6216f8b8b28d7cb27b29fd84&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR