2,6-Diaminopyridine

Warning

D24404-25G D24404-500G D24404-100G
220121-161EA 53.31 USD
220121-161 220121-162 220121-160
2,6-Diaminopyridine
2,6-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 115…118 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00006329
CAS Number: 141-86-6
EINECS: 205-507-2
UN: 2671
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR