2,4-Dinitrophenylhydrazine ≥97%

Danger

Synonyms: DNPH

D199303-25G D199303-100G D199303-500G
220121-472EA 50 USD
220121-472 220121-471 220121-473
2,4-Dinitrophenylhydrazine ≥97%
2,4-Dinitrophenylhydrazine
Formula: C₆H₆N₄O₄
MW: 198.14 g/mol
MDL Number: MFCD00007578
CAS Number: 119-26-6
EINECS: 204-309-3
UN: 3380
ADR: 4.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR