Sanger's reagent ≥99%

Warning

D1529-25ML D1529-10ML D1529-100ML
220121-112EA 440 USD
220121-112 220121-111 220121-110
Sanger's reagent ≥99%
Sanger's reagent
Formula: C₆H₃FN₂O₄
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 24 - 26 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007056
CAS Number: 70-34-8
EINECS: 200-734-3
UN: 2811
ADR: 6.1,III
Merck Index: 13,04199

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR