Acetonedimethyl acetal ≥98%, Reagent Grade

Danger

Synonyms: DMP

D136808-25ML D136808-18L D136808-500ML
220121-040EA 20.31 USD
220121-040 220121-039 220121-042
Acetonedimethyl acetal ≥98%, Reagent Grade
Acetonedimethyl acetal
Formula: C₅H₁₂O₂
MW: 104.15 g/mol
Boiling Pt: 83 °C (1013 hPa)
Melting Pt: –47 °C
Density: 0.845 g/cm³ (20 °C)
Flash Pt: –5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008479
CAS Number: 77-76-9
EINECS: 201-056-0
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR