(3,4-Dimethoxyphenyl)acetic acid ≥98%

Synonyms: Homoveratric acid, 3,4-Dimethoxyphenylacetic acid

D135909-25G D135909-100G
220121-036EA 43.08 USD
220121-036 220121-035
(3,4-Dimethoxyphenyl)acetic acid ≥98%
(3,4-Dimethoxyphenyl)acetic acid
Formula: (C₆H₃(OCH₃)₂)CH₂COOH
MW: 196.2 g/mol
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00004335
CAS Number: 93-40-3
EINECS: 202-244-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR