3',5'-Dimethoxy-4'-hydroxyacetophenone ≥97%

Synonyms: 3′,5′-Dimethoxy-4′-hydroxyacetophenone, 4'-Hydroxy-3',5'-dimethoxyacetophenone, 1-(3,5-Dimethoxy-4-hydroxyphenyl)ethan-1-one, Acetosyringone, 1-(4-Hydroxy-3,5-dimethoxyphenyl)ethanone

D134406-1G D134406-25G D134406-5G
220121-023EA 53.2 USD
220121-023 220121-024 220121-025
3',5'-Dimethoxy-4'-hydroxyacetophenone ≥97%
3',5'-Dimethoxy-4'-hydroxyacetophenone
Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Boiling Pt: 335 °C (1013 hPa)
Melting Pt: 119…120 °C
MDL Number: MFCD00008748
CAS Number: 2478-38-8
EINECS: 219-610-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR