3,5-Diaminobenzoic acid ≥98%

D12805-1KG D12805-100G
220120-458EA 628.92 USD
220120-458 220120-457
3,5-Diaminobenzoic acid ≥98%
3,5-Diaminobenzoic acid
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 546 °C (1013 hPa)
Melting Pt: 239…242 °C
Density: 1.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007807
CAS Number: 535-87-5
EINECS: 208-621-0
Merck Index: 13,02997

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR