N-Ethyldiisopropylamine ≥99%, ReagentPlus®

Danger

D125806-10L D125806-500ML
220120-439EA 1653.85 USD
220120-439 220120-440
N-Ethyldiisopropylamine ≥99%, ReagentPlus®
N-Ethyldiisopropylamine
Formula: C₈H₁₉N
Boiling Pt: 127 °C (1013 hPa)
Melting Pt: –50 °C
Density: 0.76 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00008868
CAS Number: 7087-68-5
EINECS: 230-392-0
UN: 2733
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR