trans-4-Hydroxycinnamic acid ≥98% (by HPLC)

Warning

Synonyms: p-Coumaric acid, trans-p-Cumaric Acid, trans-p-Coumaric Acid

C9008-10G C9008-1G C9008-5G C9008-25G
220117-105EA 93.66 USD
220117-105 220117-106 220117-108 220117-107
trans-4-Hydroxycinnamic acid ≥98% (by HPLC)
trans-4-Hydroxycinnamic acid
Formula: C₉H₈O₃
MW: 164.16 g/mol
MDL Number: MFCD00004399
CAS Number: 501-98-4
EINECS: 231-000-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4645cd3c9d127cab2ae6da450e82710d89ca9d8f&vid=7c075fda0e75e2d1e4bc45e5e4ab7dd0ea62103e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR