Cholesterol

Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol

C8667-1G C8667-25G C8667-100G C8667-500MG C8667-5G
220117-000EA 77.66 USD
220117-000 220117-001 220116-999 220117-002 220117-003
Cholesterol
Cholesterol
Formula: C₂₇H₄₆O
MW: 386.66 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 147 - 150 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00003646
CAS Number: 57-88-5
EINECS: 200-353-2
Merck Index: 13,02221

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR