DL-Cystine

Synonyms: (H-DL-Cys-OH)2

C8630-25G C8630-1G C8630-5G
220116-981EA 937.85 USD
220116-981 220116-980 220116-982
DL-Cystine
DL-Cystine
Formula: C₆H₁₂N₂O₄S₂
Boiling Pt: 429 °C (1013 hPa)
Melting Pt: 227 °C
MDL Number: MFCD00084652
CAS Number: 923-32-0
EINECS: 213-094-5
Merck Index: 13,02811

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR