L(-)-Cystine 98.5-101.0%

Synonyms: (R)-(-)-Cystine, (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid), (H-Cys-OH)2

C7602-25G C7602-100G C7602-10MG C7602-1KG
220116-572EA 58.58 USD
220116-572 220116-569 220116-570 220116-571
L(-)-Cystine 98.5-101.0%
L(-)-Cystine
Formula: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
MW: 240.3 g/mol
Boiling Pt: 428 °C (1013 hPa)
Melting Pt: 261…262 °C
Density: 1.677 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064228
CAS Number: 56-89-3
EINECS: 200-296-3
Merck Index: 13,02811

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR