2-Chloro-3-nitrotoluene ≥98%

C61984-1G C61984-5G
220115-559EA 56 USD
220115-559 220115-560
2-Chloro-3-nitrotoluene ≥98%
2-Chloro-3-nitrotoluene
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD00134653
CAS Number: 3970-40-9
EINECS: 223-591-9
UN: 2433
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR