Creatinine, anhydrous

Synonyms: 2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one

C4255-10MG C4255-1KG C4255-100G C4255-10G C4255-25G
220115-116EA 23.58 USD
220115-116 220115-117 220115-114 220115-115 220115-118
Creatinine, anhydrous
Creatinine
Formula: C₄H₇N₃O
MW: 113.12 g/mol
MDL Number: MFCD00059730
CAS Number: 60-27-5
EINECS: 200-466-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR