Carbol fuchsin

Danger

Synonyms: 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]-o-toluidine hydrochloride

C4165-10G C4165-25G
220115-079EA 154 USD
220115-079 220115-080
Carbol fuchsin
Carbol fuchsin
Formula: C₂₁H₂₂ClN₃
MW: 351.88 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013129
CAS Number: 4197-24-4
EINECS: 224-086-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4eed54b9ff9098b7ab5a91f46c36237e13a7a92&vid=0ba6e0569527478e56a21c9087cbb81071e3eed1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR