4-Chloro-3,5-xylenol ≥99%

Warning

Synonyms: 2-Chloro-5-hydroxy-3-xylene, Chloroxylenol, 4-Chloro-3,5-dimethylphenol

C38303-100G C38303-500G
220114-231EA 52.45 USD
220114-231 220114-232
4-Chloro-3,5-xylenol ≥99%
4-Chloro-3,5-xylenol
Formula: C₈H₉ClO
MW: 156.61 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 114…116 °C
Density: 1.364 g/cm³ (20 °C)
Flash Pt: 132 °C
MDL Number: MFCD00002324
CAS Number: 88-04-0
EINECS: 201-793-8
Merck Index: 13,02194

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR