L(+)-Cysteine hydrochloride monohydrate ≥99%

Synonyms: (R)-(+)-Cysteine hydrochloride monohydrate, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, L-Cysteine HCI Monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, Monohydrate, Cysteine Hydrochloride monohydrate

C121800-100G C121800-5G
220113-802EA 102.85 USD
220113-802 220113-803
L(+)-Cysteine hydrochloride monohydrate ≥99%
L(+)-Cysteine hydrochloride monohydrate
Formula: HSCH₂CH(NH₂)COOH·HCl·H₂O
MW: 175.64 g/mol
Melting Pt: 168 - 170 °C
Storage Temperature: Ambient
MDL Number: MFCD00065606
CAS Number: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=10395059845247050509d84df67aefcb3c98806c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR