Cyclopentanol ≥99%

C112208-5ML C112208-100ML C112208-500ML
220113-047EA 29.4 USD
220113-047 220113-045 220113-046
Cyclopentanol ≥99%
Cyclopentanol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 139 °C (1013 hPa)
Melting Pt: -19 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00001363
CAS Number: 96-41-3
EINECS: 202-504-8
UN: 2244
ADR: 3,III
Merck Index: 13,02770

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR