Tetrabromomethane ≥99%, ReagentPlus®

Warning

Synonyms: Carbon tetrabromide

C11081-100G C11081-5G C11081-500G
220113-163EA 84.75 USD
220113-163 220113-165 220113-164
Tetrabromomethane ≥99%, ReagentPlus®
Tetrabromomethane
Formula: Br₄C
MW: 331.63 g/mol
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: 90 - 92 °C
Density: 3.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000117
CAS Number: 558-13-4
EINECS: 209-189-6
UN: 2516
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR