Cyclohexylacetic acid

Synonyms: Cyclohexaneacetic acid

C104507-5G C104507-25G
220113-585EA 54.71 USD
220113-585 220113-584
Cyclohexylacetic acid
Cyclohexylacetic acid
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: 26 - 27 °C
Density: 0.9985 g/cm³ (22 °C)
MDL Number: MFCD00001518
CAS Number: 5292-21-7
EINECS: 226-132-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR