1,2-Epoxycyclohexane ≥98%

Danger

Synonyms: Epoxycyclohexane, Cyclohexene oxide

C102504-25ML C102504-500ML C102504-100ML
220113-565EA 62.37 USD
220113-565 220113-566 220113-564
1,2-Epoxycyclohexane ≥98%
1,2-Epoxycyclohexane
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 129 °C (1013 hPa)
Melting Pt: –27 °C
Density: 0.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005162
CAS Number: 286-20-4
EINECS: 206-007-7
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR