Crystal violet, certified by the Biological Stain Commission

Danger

Synonyms: CI 42555

C0775-25G C0775-100G
220113-131EA 92.68 USD
220113-131 220113-130
Crystal violet, certified by the Biological Stain Commission
Crystal violet
Formula: C₂₅H₃₀ClN₃
MW: 407.99 g/mol
Melting Pt: 189 - 194 °C
Density: 1.19 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011750
CAS Number: 548-62-9
EINECS: 208-953-6
UN: 3077
ADR: 9,III
Merck Index: 13,04406
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3952a823a2ebb8660f8e92b107344bdfddd4747c&vid=a88f2b02bb42c167ddd8b868c13962e60d28414c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR