Caffeic acid

Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid

C0625-25G C0625-2G C0625-5G
220113-027EA 249.23 USD
220113-027 220113-028 220113-029
Caffeic acid
Caffeic acid
Formula: C₉H₈O₄
MW: 180.16 g/mol
Boiling Pt: 366 °C (1013 hPa)
Melting Pt: 234…237 °C
MDL Number: MFCD00004392
CAS Number: 331-39-5
EINECS: 206-361-2
Merck Index: 12,01673

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR