tert-Butylamine ≥98%

Danger

Synonyms: 2-Aminoisobutane

B89205-1L B89205-5ML B89205-500ML B89205-100ML
220112-752EA 72.48 USD
220112-752 220112-754 220112-753 220112-751
tert-Butylamine ≥98%
tert-Butylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 44.4 °C (1013 hPa)
Melting Pt: –72 °C
Density: 0.6965 g/cm³ (20 °C)
Flash Pt: –38 °C
Storage Temperature: Ambient
MDL Number: MFCD00008050
CAS Number: 75-64-9
EINECS: 200-888-1
UN: 2734
ADR: 8,I
Merck Index: 13,01544

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR